Silver sulphate 6,6 g/l in sulphuric acid, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Disilver sulphate
30492.241EA 75 EUR
30492.241 30492.423
Silver sulphate 6,6 g/l in sulphuric acid, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Silver sulphate
Formule: Ag₂SO₄
MW: 311.8 g/mol
Kookpunt: 1085
Smeltpunt: 655
MDL Nummer: MFCD00003407
CAS nummer: 10294-26-5
EINECS: 233-653-7
UN: 1830
ADR: 8 II
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Chemical oxygen demand (COD) Passes test
NIST traceable Confirmed