Kaliumhydroxide 50% (w/v) in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Potash caustic, Caustic potash
35621.AP 35621.K2
35621.APEA 52.2 EUR
35621.AP 35621.K2
Kaliumhydroxide 50% (w/v) in aqueous solution
Kaliumhydroxide
CAS No.: 1310-58-3 Molecular Formula: HKO Formula Weight: 56.11 Physical Form: Liquid Density: 1.50 Appearance: Colourless UN No.: 1814 MDL No.: MFCD00003553
Formule: KOH
MW: 56.11 g/mol
Kookpunt: 1320
Smeltpunt: 360
Dichtheid: 1.50
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806