Tetrabutylammonium hydroxide 40% (w/w) in methanol
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
366321000 366325000 366320025
ACRO366321000EA 81.2 EUR
ACRO366321000 ACRO366325000 ACRO366320025
Tetrabutylammonium hydroxide 40% (w/w) in methanol
Tetrabutylammonium hydroxide
Formule: C₁₆H₃₇NO
MW: 259.48 g/mol
Smeltpunt: –98 °C
Dichtheid: 0.91 g/cm³
Vlampunt: 12 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009425
CAS nummer: 2052-49-5
EINECS: 218-147-6
UN: 3286
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear to slightly hazy colorless to light yellow Solution
Titration with HClO4 ≥40 %