Tetrabutylammonium hydroxide 40% in aqueous solution
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
5026.1 5026.2
ROTH5026.1EA 0 EUR
ROTH5026.1 ROTH5026.2
Tetrabutylammonium hydroxide 40% in aqueous solution
Tetrabutylammonium hydroxide
Formule: C₁₆H₃₇NO
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009425
CAS nummer: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8,II