Tetramethylammonium hydroxide 25% (w/w) in aqueous solution , 99,9999% (metals basis), Electronic Grade
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 99.9999%
CAS No.: 75-59-2
Molecular Formula: C4H13NO
Formula Weight: 91.15
Physical Form: Liquid
Density: 1.016
Refractive Index: 1.3840
Sensitivity: Air Sensitive
Appearance: Colourless to pale yellow
Boiling Point: ca 102°
Beilstein Reference: 3558708
Merck Reference: 14,9224
UN No.: 1835
MDL No.: MFCD00008280
Formule:
C₄H₁₃NO Kookpunt: ∼102 °C Dichtheid: 1,016 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008280 CAS nummer: 75-59-2 EINECS: 200-882-9 UN: 1835 ADR: 8,II Merck Index: 14,9224 |