Ammoniak-oplossing 25% ≥99.99% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Ammoniumhydroxide, Waterig-ammoniakale oplossing
87903.AK 87903.AU
87903.AKEA 138 EUR
87903.AK 87903.AU
Ammoniak-oplossing 25% ≥99.99% (metals basis)
Ammoniak-oplossing
Purity: 99.99% CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Physical Form: Liquid Density: 0.91 Appearance: Colourless Odour: Ammonia-like pH: 12 Merck Reference: 14,494 UN No.: 2672 MDL No.: MFCD00066650
Formule: NH₃
MW: 35.05 g/mol
Kookpunt: 32…37.7 °C (1013 hPa)
Smeltpunt: –60…–57.5 °C
Dichtheid: 0.91
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00066650
CAS nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III
Merck Index: 14,494