PCR/gel clean-up kits
Leverancier: ENZO LIFE SCIENCES
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ENZGEN1000020
ENZGEN1000050
ENZGEN1000200
ENZOENZGEN1000020EA
66
EUR
ENZOENZGEN1000020
ENZOENZGEN1000050
ENZOENZGEN1000200
PCR/gel clean-up kits
Reagentia voor nucleïnezuren
Nucleic Acid Purification Kits and Reagents
PCR/Gel Clean Up Kits are quick and simple kits for purifying DNA from PCR, gels and labelling reactions for a variety of downstream applications.
- High recoveries for fragments down to 50 bp
- Small elution volume down to 15 µl
- Binding buffer with pH indicator
- Validated for use with CGH labelling kit
- Less preparation time: 10 mins for 6 preparations
Clean-up columns use silica membrane technology in a mini spin column format to deliver easy and efficient DNA purification from PCR products, agarose gels, and labelling reactions.