Ammonium sulphide 40 - 44% (w/w) in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
40509.AE 40509.AP
40509.AEEA 92.1 EUR
40509.AE 40509.AP
Ammonium sulphide 40 - 44% (w/w) in aqueous solution
Ammonium sulphide
CAS No.: 12135-76-1 Molecular Formula: (NH4)2S Formula Weight: 68.14 Physical Form: Liquid Appearance: Yellow to orange Odour: Stench UN No.: 2683 MDL No.: MFCD00010892
Formule: H₈N₂S
MW: 68.14 g/mol
Smeltpunt: -18
Dichtheid: 0.99
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010892
CAS nummer: 12135-76-1
EINECS: 235-223-4
UN: 2683
ADR: 8 II
Merck Index: 13,00560