Hydrazine hydrate 80% ≥80% for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Hydrazinium hydroxide, Hydrazine monohydrate
M-Clarity™ quality level = MQ200
8.04604.9055 8.04604.0250 8.04604.1000
8.04604.9055EA 0 EUR
8.04604.9055 MERC8.04604.0250 MERC8.04604.1000
Hydrazine hydrate 80% ≥80% for synthesis, Sigma-Aldrich®
Hydrazine hydrate
Formule: NH₂NH₂·xH₂O
MW: 32.05 g/mol (anhy)
Dichtheid: 1.02 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149931
CAS nummer: 10217-52-4
EINECS: 206-114-9
UN: 2030
ADR: 8 (6.1) II