Ethidiumbromide 10 mg/ml aqueous solution
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
2218.1 2218.2
ROTH2218.1EA 0 EUR
ROTH2218.1 ROTH2218.2
Ethidiumbromide 10 mg/ml aqueous solution
Ethidiumbromide
Formule: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Kookpunt: 72
MDL Nummer: MFCD00011724
CAS nummer: 1239-45-8
EINECS: 214-984-6
Merck Index: 13,04751