L(+)-Cysteine hydrochloride monohydrate
Leverancier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
03388-1 03388-2
BMOL03388-1EA 0 EUR
BMOL03388-1 BMOL03388-2
L(+)-Cysteine hydrochloride monohydrate
L(+)-Cysteine hydrochloride monohydrate
Formule: HSCH₂CH(NH₂)COOH·HCl·H₂O
Smeltpunt: 168…170 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00065606
CAS nummer: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=10395059845247050509d84df67aefcb3c98806c&prid=web&clid=SAPDAM