Kaliumhydroxide concentrate for dilution to 1L, 0.1 M KOH (0.1N) volumetric solution, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Potash caustic, Caustic potash
38070-1EA 38070-6X1EA
38070-1EAEA 51 EUR
38070-1EA 38070-6X1EA
Kaliumhydroxide concentrate for dilution to 1L, 0.1 M KOH (0.1N) volumetric solution, Fluka™
Kaliumhydroxide
Concentrate for 1L standard solution
Formule: KOH
MW: 56.11 g/mol
Kookpunt: >100 °C (1.013 hPa)
Smeltpunt: 360
Dichtheid: 1.140 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806
Order Now
Specification Test Results
Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective