Natriumhydroxide concentrate for dilution to 0.5 N in aqueous solution, Acculute™ standard volumetric solution
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
88620.LQ 88620.LS
88620.LQEA 39.4 EUR
88620.LQ 88620.LS
Natriumhydroxide concentrate for dilution to 0.5 N in aqueous solution, Acculute™ standard volumetric solution
Natriumhydroxide
CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid Odour: Odourless UN No.: 1824 MDL No.: MFCD00003548
Formule: NaOH
Kookpunt: ∼105 °C (1013 hPa)
Dichtheid: 1,327 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003548
CAS nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II