Lysates, HeLa (human)
Leverancier: ENZO LIFE SCIENCES
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ADILYCHL100F
ADILYCHL101F
ADILYCHL102F
ENZOADILYCHL100FEA
165
EUR
ENZOADILYCHL100F
ENZOADILYCHL101F
ENZOADILYCHL102F
Lysates, HeLa (human)
Cellen
The HeLa cell line derived from a human donor's epithelioid carcinoma tissue (cervix).
Dilution range: 20 µg/lane (Western blotting)
Liquid lysates in SDS-PAGE sample buffer.
Heat shock-treated HeLa cells with no recovery time can be a useful tool in studying the regulators of immediate-early gene expression during heat shock stress response.