Guanidinium thiocyanate
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
39577.01
39577.02
SERA39577.01EA
71
EUR
SERA39577.01
SERA39577.02
Guanidinium thiocyanate
Guanidinium thiocyanate
Formule:
CH₅N₃·CHNS Smeltpunt: 118 °C Dichtheid: 1,29 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00013027 CAS nummer: 593-84-0 EINECS: 209-812-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...