Calcium oxalate monohydrate ≥99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Oxalic acid calcium salt monohydrate, Calcium ethanedioate monohydrate
32430.22
32430.36
32430.22EA
47
EUR
32430.22
32430.36
Calcium oxalate monohydrate ≥99%
Calcium oxalate monohydrate
Purity: 99%
CAS No.: 5794-28-5
Molecular Formula: CaC2O4.H2O
Formula Weight: 146.11 (128.10anhy)
Physical Form: Powder/lump
Density: 2.12
Melting Point: 200°
Merck Reference: 14,1685
UN No.: 3288
MDL No.: MFCD00150039
Formule:
C₂CaO₄·1H₂O Smeltpunt: 200 °C Dichtheid: 2,12 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00150039 CAS nummer: 5794-28-5 EINECS: 209-260-1 UN: 3288 ADR: 6.1,III Merck Index: 14,1685 |