di-Ammonium oxalate monohydrate ≥99.0%, powder ACS
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate
0219140891 0219140880 0219140890
ICNA0219140891EA 161 EUR
ICNA0219140891 ICNA0219140880 ICNA0219140890
di-Ammonium oxalate monohydrate ≥99.0%, powder ACS
di-Ammonium oxalate monohydrate
Chloride ≤0,002% Iron ≤2 ppm sulphate ≤0,002%
Formule: (NH₄)₂C₂O₄·H₂O
Kookpunt: 70 °C (1013 hPa)
Smeltpunt: 70 °C
Dichtheid: ∼1,5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149694
CAS nummer: 6009-70-7
EINECS: 214-202-3
UN: 3288
ADR: 6.1,III
Merck Index: 13,00541
Order Now
Specification Test Results
Synonyms Ethanedioic Acid Monohydrate; Oxalic acid diammonium salt
Form White Powder
Storage Store at room temperature (15 to 30 °C)
Heavy Metals ≤5 ppm
Insoluble Matter ≤0,005%
Residue on Ignition ≤0,02%
Chloride ≤0.002%
Iron ≤2 ppm
Sulphate ≤0.002%