tri-Sodium citrate dihydrate
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
38642.01
38642.02
38642.03
SERA38642.01EA
0
EUR
SERA38642.01
SERA38642.02
SERA38642.03
tri-Sodium citrate dihydrate
tri-Sodium citrate dihydrate
Formule:
Na₃C₆H₅O₇·2H₂O Smeltpunt: 150 °C Dichtheid: 1,76 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00150031 CAS nummer: 6132-04-3 EINECS: 200-675-3 Merck Index: 13,08675 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...