EDTA disodium salt dihydrate 99.0-101.0% ACS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate
33312.22 33312.36
33312.22EA 36.2 EUR
33312.22 33312.36
EDTA disodium salt dihydrate 99.0-101.0% ACS
EDTA disodium salt dihydrate
Purity: 99.0-101.0% CAS No.: 6381-92-6 Molecular Formula: (NaO2CCH2)2NCH2CH2N(CH2CO2H)2.2H2O Formula Weight: 372.24 (336.22anhy) Physical Form: Powder Melting Point: 248° dec. Beilstein Reference: 3900609 MDL No.: MFCD00150037
Formule: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Smeltpunt: 248 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150037
CAS nummer: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM