EDTA disodium salt dihydrate
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate
8043.1 8043.2 8043.3 8043.4 8043.5
ROTH8043.1EA 0 EUR
ROTH8043.1 ROTH8043.2 ROTH8043.3 ROTH8043.4 ROTH8043.5
EDTA disodium salt dihydrate
EDTA disodium salt dihydrate
Formule: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Smeltpunt: 255
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150037
CAS nummer: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM