Potassium hexacyanoferrate(II) trihydrate 98
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Potassiumhexacyanoferrate(II) trihydrate, Tetrapotassium hexacyanoferrate trihydrate, Potassium ferrocyanide trihydrate
A15736.0E A15736.22 A15736.36
A15736.0EEA 220 EUR
A15736.0E A15736.22 A15736.36
Potassium hexacyanoferrate(II) trihydrate 98
Potassium hexacyanoferrate(II) trihydrate
Purity: 98+% CAS No.: 14459-95-1 Molecular Formula: K4Fe(CN)6.3H2O Formula Weight: 422.39 (368.36anhy) Odour: Odourless pH: 9.5 Melting Point: 69-71° Merck Reference: 14,7631 MDL No.: MFCD00167023
Formule: K₄Fe(CN)₆·3H₂O
MW: 422.39 g/mol
Smeltpunt: 69…71 °C
Dichtheid: 1.85
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00167023
CAS nummer: 14459-95-1
EINECS: 237-722-2
Merck Index: 12,07793