Potassium hexacyanoferrate(II) trihydrate, extra_pure, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Potassiumhexacyanoferrate(II) trihydrate, Tetrapotassium hexacyanoferrate trihydrate, Potassium ferrocyanide trihydrate
MQ200
1.04982.1000 1.04982.9025
1.04982.1000EA 190 EUR
1.04982.1000 1.04982.9025
Potassium hexacyanoferrate(II) trihydrate, extra_pure, Supelco®
Potassium hexacyanoferrate(II) trihydrate
Formule: K₄Fe(CN)₆·3H₂O
MW: 422.39 g/mol
Smeltpunt: 70
Dichtheid: 1.889 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00167023
CAS nummer: 14459-95-1
EINECS: 237-722-2
Merck Index: 12,07793