Sodium molybdate(VI) dihydrate
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate
0274.1 0274.2 0274.3
ROTH0274.1EA 0 EUR
ROTH0274.1 ROTH0274.2 ROTH0274.3
Sodium molybdate(VI) dihydrate
Sodium molybdate(VI) dihydrate
Formule: Na₂MoO₄·2H₂O
MW: 241.95 g/mol
Smeltpunt: 687
Dichtheid: 3.28 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149170
CAS nummer: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718