Chromium(III) potassium sulphate dodecahydrate 98.0-102.0% ACS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Chromium potassium sulphate dodecahydrate, Chromium (III) potassium bis(sulphate) dodecahydrate
36715.22 36715.36
36715.22EA 38.1 EUR
36715.22 36715.36
Chromium(III) potassium sulphate dodecahydrate 98.0-102.0% ACS
Chromium(III) potassium sulphate dodecahydrate
Purity: 98.0-102.0% CAS No.: 7788-99-0 Molecular Formula: CrKO8S2.12H2O Formula Weight: 499.39 (283.21anhy) Physical Form: Solid Density: 1.83 Appearance: Dark purple Odour: Odourless Melting Point: 89° Merck Reference: 14,2229 MDL No.: MFCD00149917
Formule: CrK(SO₄)₂·12H₂O
MW: 499.4 g/mol
Smeltpunt: 89
Dichtheid: 1.83
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149917
CAS nummer: 7788-99-0
EINECS: 233-401-6
Merck Index: 14,2229