Magnesium perchlorate hexahydrate ≥99% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
11635.30 11635.A1
11635.30EA 61.7 EUR
11635.30 11635.A1
Magnesium perchlorate hexahydrate ≥99% (metals basis)
Magnesium perchlorate hexahydrate
Purity: 99% CAS No.: 13446-19-0 Molecular Formula: Mg(ClO4)2.6H2O Formula Weight: 331.33 (223.23anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,5678 UN No.: 1475 MDL No.: MFCD00149778
Formule: Cl₂MgO₈·6H₂O
MW: 331.3 g/mol
Smeltpunt: 185…190 °C
Dichtheid: 1.98 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149778
CAS nummer: 13446-19-0
EINECS: 233-108-3
UN: 1475
ADR: 5.1 II
Merck Index: 14,5678