Barium hydroxide octahydrate ≥97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
A12714.0C A12714.0E A12714.36
A12714.0CEA 342 EUR
A12714.0C A12714.0E A12714.36
Barium hydroxide octahydrate ≥97%
Barium hydroxide octahydrate
Purity: 97% CAS No.: 12230-71-6 Molecular Formula: H2BaO2.8H2O Formula Weight: 315.48 (171.36anhy) Odour: Odourless Melting Point: 78° UN No.: 3262 MDL No.: MFCD00149152
Formule: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Kookpunt: 780
Smeltpunt: 78
Dichtheid: 2.18
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149152
CAS nummer: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8 III
Merck Index: 13,00980