Calcium sulphate dihydrate
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Plaster of Paris
0256.1
0256.2
0256.3
ROTH0256.1EA
0
EUR
ROTH0256.1
ROTH0256.2
ROTH0256.3
Calcium sulphate dihydrate
Calcium sulphate dihydrate
Formule:
CaSO₄·2H₂O MW: 172.17 g/mol Dichtheid: 2.32 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00149625 CAS nummer: 10101-41-4 EINECS: 231-900-3 Merck Index: 13,01711 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...