Magnesium sulphate heptahydrate
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt
P027.1 P027.2 P027.3
ROTHP027.1EA 30.9 EUR
ROTHP027.1 ROTHP027.2 ROTHP027.3
Magnesium sulphate heptahydrate
Magnesium sulphate heptahydrate
Formule: MgSO₄·7H₂O
MW: 246.48 g/mol
Smeltpunt: 1124
Dichtheid: 2.66 g/cm³ (20 °C)
MDL Nummer: MFCD00149785
CAS nummer: 10034-99-8
Merck Index: 13,05715