Aluminium chloride hexahydrate, Reagent Grade
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Aluminum chloride hexahydrate
12297.36 12297.A3
12297.36EA 55.1 EUR
12297.36 12297.A3
Aluminium chloride hexahydrate, Reagent Grade
Aluminium chloride hexahydrate
Pharmaceuticals, pigments, in cosmetics and deOdourants, dyeing fabrics, refining crude oils, parchment paper manufacturing
Formule: AlCl₃·6H₂O
MW: 241.43 g/mol
Smeltpunt: 100 °C (ontbinding)
Dichtheid: 2.398
MDL Nummer: MFCD00149134
CAS nummer: 7784-13-6
UN: 3260
ADR: 8 III
Merck Index: 14,337