Aluminium chloride hexahydrate 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Aluminum chloride hexahydrate
A14437.0B A14437.0I A14437.30
A14437.0BEA 48.6 EUR
A14437.0B A14437.0I A14437.30
Aluminium chloride hexahydrate 99%
Aluminium chloride hexahydrate
Formule: AlCl₃·6H₂O
MW: 241.43 g/mol
Smeltpunt: 100 °C (ontbinding)
Dichtheid: 2.398
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149134
CAS nummer: 7784-13-6
EINECS: 231-208-1
UN: 3260
ADR: 8 III