Aluminium nitrate nonahydrate ≥99.999% (metals basis), Puratronic® low in mercury
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Aluminum nitrate nonahydrate
43145.22 43145.14 43145.36
43145.22EA 374 EUR
43145.22 43145.14 43145.36
Aluminium nitrate nonahydrate ≥99.999% (metals basis), Puratronic® low in mercury
Aluminium nitrate nonahydrate
Purity: 99.999% CAS No.: 7784-27-2 Molecular Formula: Al(NO3)3.9H2O Formula Weight: 375.13 (212.99anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,352 UN No.: 1438 MDL No.: MFCD00003419
Formule: Al(NO₃)₃·9H₂O
MW: 375.13 g/mol
Smeltpunt: 73.5 °C
Dichtheid: 1.72 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149132
CAS nummer: 7784-27-2
EINECS: 236-751-8
UN: 1438
ADR: 5.1 III
Merck Index: 14,352