Bole white (kaolin), pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
211740010 211740025 211740100
ACRO211740010EA 41.3 EUR
ACRO211740010 ACRO211740025 ACRO211740100
Bole white (kaolin), pure
Bole white (kaolin)
Formule: Al₂SiO₇·2H₂O
Smeltpunt: 1750 °C
Dichtheid: 0,5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00062311
CAS nummer: 1332-58-7
EINECS: 310-194-1
Bereik: exempt
Merck Index: 13,05300
Order Now
Specification Test Results
Appearance Light beige Powder
Loss on drying ≤1.5 %
Particle size 1.0 to 1.8 µm (D50)
Loss on ignition 10.8 % to 11.8 %