Ammonium iron(II) sulphate hexahydrate
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate
P728.1 P728.2
ROTHP728.1EA 34.9 EUR
ROTHP728.1 ROTHP728.2
Ammonium iron(II) sulphate hexahydrate
Ammonium iron(II) sulphate hexahydrate
Formule: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Smeltpunt: 100
Dichtheid: 1.864 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150530
CAS nummer: 7783-85-9
EINECS: 233-151-8