Cerium trichloride heptahydrate ≥99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Cerous chloride heptahydrate, Cerium (III) chloride heptahydrate
A12947.0B A12947.18 A12947.30
A12947.0BEA 413 EUR
A12947.0B A12947.18 A12947.30
Cerium trichloride heptahydrate ≥99%
Cerium trichloride heptahydrate
Purity: 99% CAS No.: 18618-55-8 Molecular Formula: CeCl3.7H2O Formula Weight: 372.60 (246.48anhy) Odour: Odourless Merck Reference: 14,1997 MDL No.: MFCD00149634
Formule: CeCl₃·7H₂O
MW: 372.58 g/mol
Smeltpunt: 848
Dichtheid: 3.97 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149634
CAS nummer: 18618-55-8
EINECS: 606-073-0
Merck Index: 12,02041