Ytterbium(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
11193.09 11193.18
11193.09EA 59.9 EUR
11193.09 11193.18
Ytterbium(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Ytterbium(III) chloride hydrate
Purity: 99.9% CAS No.: 19423-87-1 Molecular Formula: YbCl3.xH2O (x≈6) Formula Weight: 279.40(anhy) Physical Form: Wet Crystalline Aggregates Density: 2.58 Appearance: Green Odour: Odourless Melting Point: 150-155° Merck Reference: 14,10105 MDL No.: MFCD00149885
Formule: Cl₃Yb·H₂O
MW: 297.41 g/mol
Smeltpunt: 150…155 °C
Dichtheid: 2.58
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149885
CAS nummer: 19423-87-1
EINECS: 233-800-5
Merck Index: 14,10105