Sandbaths for modular heating blocks
Leverancier: OHAUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30400175 30400176 30400173 30400174
OHAU30400175EA 330 EUR
OHAU30400175 OHAU30400176 OHAU30400173 OHAU30400174
Sandbaths for modular heating blocks
Verwarmers Zandbaden
Ideal for irregular shaped vessels.
  • Stainless steel construction for superior corrosion resistance
  • Designed to hold sand, stainless steel shot or non-volatile liquids