Low temperature covers
Leverancier: OHAUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30400179 30400180 30400181
OHAU30400179EA 121 EUR
OHAU30400179 OHAU30400180 OHAU30400181
Low temperature covers
Blokverwarmers Blokken
Plexiglass cover reduces air flow for additional temperature stability in low temperature applications.
  • Two sides are open 6,4 mm