Copper(II) nitrate hemipentahydrate ≥98% (≥26% Cu basis), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Cupric Nitrate Hydrate, Copper dinitrate hemipentahydrate, Cupric nitrate hemi(pentahydrate), Cupric nitrate 2.5-hydrate, Cupric nitrate hemipentahydrate
12837-1KG 12837-2.5KG 12837-500G
12837-1KGEA 110 EUR
12837-1KG 12837-2.5KG 12837-500G
Copper(II) nitrate hemipentahydrate ≥98% (≥26% Cu basis), Fluka™
Copper(II) nitrate hemipentahydrate
Formule: Cu(NO₃)₂·2.5H₂O
Smeltpunt: 114 °C
Dichtheid: 2,320 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02253001
CAS nummer: 19004-19-4
EINECS: 221-838-5
UN: 3085
ADR: 5.1,I
Merck Index: 13,02671
Order Now
Specification Test Results
Assay Min. 98 %
Assay of Cu Min. 26 %
Water insoluble matter Max. 0.003 %
Iron (Fe) Max. 0.005 %
Chloride (Cl) Max. 0.005 %
Sulfate (SO₄) Max. 0.01 %