Koper(II)sulfaat pentahydraat, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, extra pure suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Cupric sulphate pentahydrate, Copper sulphate pentahydrate
1.02787.1000 1.02787.5000 1.02787.9050
1.02787.1000EA 405 EUR
1.02787.1000 1.02787.5000 1.02787.9050
Koper(II)sulfaat pentahydraat, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, extra pure suitable for use as excipient, SAFC®
Koper(II)sulfaat pentahydraat
Formule: CuSO₄·5H₂O
MW: 249.69 g/mol
Dichtheid: 2.284 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149681
CAS nummer: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9 III
Bereik: 01-2119520566-40
Merck Index: 13,02682