Scandium(III) chloride hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11219.03
11219.06
11219.03EA
201
EUR
11219.03
11219.06
Scandium(III) chloride hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Scandium(III) chloride hexahydrate
Purity: 99.99%
CAS No.: 20662-14-0
Molecular Formula: ScCl3.6H2O
Formula Weight: 259.41 (151.32anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Merck Reference: 14,8392
MDL No.: MFCD00149852
Formule:
Cl₃Sc·6H₂O MW: 259.41 g/mol Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00149852 CAS nummer: 20662-14-0 EINECS: 233-799-1 Merck Index: 14,8392 |