Chromium(III) nitrate nonahydrate ≥99.99% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Chromium trinitrate nonahydrate, Chromic nitrate nonahydrate, Chromium nitrate nonahydrate
43182.22 43182.14 43182.06
43182.22EA 674 EUR
43182.22 43182.14 43182.06
Chromium(III) nitrate nonahydrate ≥99.99% (metals basis)
Chromium(III) nitrate nonahydrate
Purity: 99.99% min CAS No.: 7789-02-8 Molecular Formula: Cr(NO3)3.9H2O Formula Weight: 400.14 (238.01anhy) Physical Form: Crystalline Appearance: Purple Odour: Odourless Melting Point: 60° Merck Reference: 14,2226 UN No.: 2720 MDL No.: MFCD00149659
Formule: CrN₃O₉·9H₂O
MW: 400.15 g/mol
Smeltpunt: 60
Dichtheid: 1.807 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149659
CAS nummer: 7789-02-8
EINECS: 236-921-1
UN: 2720
ADR: 5.1 III
Merck Index: 14,2226