Manganese(II) sulphate monohydrate
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Manganese sulphate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate
4487.1
4487.2
ROTH4487.1EA
0
EUR
ROTH4487.1
ROTH4487.2
Manganese(II) sulphate monohydrate
Manganese(II) sulphate monohydrate
Formule:
MnSO₄·1H₂O Smeltpunt: 700 °C Dichtheid: 2,95 g/cm³ (25 °C) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00149159 CAS nummer: 10034-96-5 EINECS: 232-089-9 UN: 3077 ADR: 9,III Merck Index: 13,05763 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...