Cobalt(II) chloride hexahydrate, pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
192860010 192862500
ACRO192860010EA 251 EUR
ACRO192860010 ACRO192862500
Cobalt(II) chloride hexahydrate, pure
Cobalt(II) chloride hexahydrate
Formule: CoCl₂·6H₂O
Kookpunt: 110 °C (1013 hPa)
Smeltpunt: 86 °C
Dichtheid: 1,92 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149652
CAS nummer: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 13,02462
Order Now
Specification Test Results
Appearance Red to violet Crystals
Titration Complexometric 98.0 to 102.0 %