Cobalt(II) chloride hexahydrate
Leverancier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate
1.02533.1000
1.02533.9025
1.02533.1000EA
524
EUR
1.02533.1000
1.02533.9025
Cobalt(II) chloride hexahydrate
Cobalt(II) chloride hexahydrate
Formule:
CoCl₂·6H₂O MW: 237.93 g/mol Kookpunt: 110 Smeltpunt: 56 Dichtheid: 1.92 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00149652 CAS nummer: 7791-13-1 EINECS: 231-589-4 UN: 3077 ADR: 9 III Merck Index: 13,02462 |