Kobalt(II)nitraat hexahydraat ≥97.7%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Cobalt dinitrate hexahydrate, Cobaltous nitrate hexahydrate
11341.14
11341.30
11341.A1
11341.A7
11341.14EA
59.2
EUR
11341.14
11341.30
11341.A1
11341.A7
Kobalt(II)nitraat hexahydraat ≥97.7%
Cobalt(II) nitrate hexahydrate
Purity: 97.7% min
CAS No.: 10026-22-9
Molecular Formula: Co(NO3)2.6H2O
Formula Weight: 291.04 (182.94anhy)
Physical Form: Crystalline
Density: 1.87
Refractive Index: 1.52
Appearance: Red
Odour: Odourless
Melting Point: 55-56°
Merck Reference: 14,2444
UN No.: 1477
MDL No.: MFCD00149647
Formule:
Co(NO₃)₂·6H₂O MW: 291.03 g/mol Smeltpunt: 55…56 °C Dichtheid: 1.87 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00149647 CAS nummer: 10026-22-9 EINECS: 233-402-1 UN: 1477 Merck Index: 13,02469 |