L(+)-Glutamine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4030662.0100 4030662.0500
BACE4030662.0100EA 0 EUR
BACE4030662.0100 BACE4030662.0500
L(+)-Glutamine
L(+)-Glutamine
Formule: C₅H₁₀N₂O₃
Kookpunt: 355 °C (1013 hPa)
Smeltpunt: 185…186 °C
Dichtheid: 1,5326 g/cm³ (130 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008044
CAS nummer: 56-85-9
EINECS: 200-292-1
Merck Index: 13,04484
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=15d7d87daff0fec33a974a578415e157a3278049&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR