Penicillin : Streptomycin solution_SC 6,0/10,0 g/L 100X
Leverancier: BIOWEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L0022-100 L0022-020
L0022-100EA 17.5 EUR
L0022-100 L0022-020
Penicillin : Streptomycin solution_SC 6,0/10,0 g/L 100X
Penicillin/Streptomycin
Penicillin-Streptomycin acts against Gram-negative and Gram-positive bacteria.
  • Store at –20 °C
  • pH of 6±1
  • Shelf life of 18 months
Endotoxin level at 10 ml/l of solution is <0,1 EU/ml.
Opgelet: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.
CAS nummer: 1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)