L(+)-Glutamine, EMPROVE® EXPERT DAB, USP suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
1.00286.1000 1.00286.9010
1.00286.1000EA 844 EUR
1.00286.1000 1.00286.9010
L(+)-Glutamine, EMPROVE® EXPERT DAB, USP suitable for use as excipient, SAFC®
L(+)-Glutamine
Formule: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Kookpunt: 355
Smeltpunt: 185…186 °C
Dichtheid: 1.5326 g/cm³ (130 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008044
CAS nummer: 56-85-9
EINECS: 200-292-1
Merck Index: 13,04484
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=15d7d87daff0fec33a974a578415e157a3278049&prid=web&clid=SAPDAM