Endocannabinoid library plates, SCREEN-WELL®
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BML28010100 BML28010500
ENZOBML28010100EA 2950 EUR
ENZOBML28010100 ENZOBML28010500
Endocannabinoid library plates, SCREEN-WELL®
Microplaten HTS-microplaten
SCREEN-WELL® Endocannabinoid Library contains 58 compounds with defined, putative, potential or speculative activity at cannabinoid (CB) and vanilloid (VR) receptors.
The library is an array of 10 different fatty acids and 6 different polar head groups. Ligands are supplied dissolved in biocompatible solvents at 1 mM and aliquoted into deep-well plates. The library is a rich source of ligands for receptor de-orphaning and includes the following classes of ligands: Amides, ethanolamides, lipo-amino acids, acyl-GABAs, acyl-dopamines.