Fatty acid library plates, SCREEN-WELL®
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BML28030100 BML28030500
ENZOBML28030100EA 1060 EUR
ENZOBML28030100 ENZOBML28030500
Fatty acid library plates, SCREEN-WELL®
Microplaten HTS-microplaten
SCREEN-WELL® Fatty Acid Library contains 64 fatty acids of varying saturation and chain length.
A diverse set of fatty acids can be a rich source of compounds for receptor de-orphaning, inhibitor screening, and high content screening. This set contains both fatty acids with defined activities and fatty acids with putative biological activities.Library includes: Unsaturated & saturated fatty acids, arachidonic acid, C10-24 chain length linolenic acid, E and Z isomers, γ-linolenic acid, and many more.
Fatty acids are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates. Concentration of 10 mM in DMSO.